Learn More
Medchemexpress LLC (S,R,S)-AHPC-O-Ph-PEG1-NH2 | 2361117-24-8 | 97.0% | 50 MG
SDP
Catalog No. 5000338941
Encompass_Preferred
(S,R,S)-AHPC-O-Ph-PEG1-NH2 is an E3 ligase ligand-linker conjugate that incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker. It is utilized in PROTAC EED degrader-1 (HY-130614), which is a PROTAC targeting EED with a pKD of 9.02.
Catalog No. 50-003-38941 Supplier Medchemexpress LLC Supplier No. HY13081650MG
(S,R,S)-AHPC-O-Ph-PEG1-NH2 is an E3 ligase ligand-linker conjugate that incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker. It is utilized in PROTAC EED degrader-1 (HY-130614), which is a PROTAC targeting EED with a pKD of 9.02.