Learn More
TARGETMOL CHEMICALS INC S R S-AHPC-PEG1-N3 10MG
SDP
Catalog No. 502876900
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (S,R,S)-AHPC-PEG1-N3 (E3 ligase Ligand-Linker Conjugates 3) is a synthesized E3 ligase ligand-linker conjugate that includes the (S,R,S)-AHPC-based VHL ligand and a 1-unit PEG linker used in PROTAC technology.
Catalog No. 50-287-6900 Supplier TARGETMOL CHEMICALS INC Supplier No. T1792010MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (S,R,S)-AHPC-PEG1-N3 (E3 ligase Ligand-Linker Conjugates 3) is a synthesized E3 ligase ligand-linker conjugate that includes the (S,R,S)-AHPC-based VHL ligand and a 1-unit PEG linker used in PROTAC technology.