Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-PEG1-N3 | 2101200-09-1 | 98.8% | 557.67 g/mol | C26H35N7O5S | 1 G
SDP

Catalog No. 5000244952
Encompass_Preferred

(S,R,S)-AHPC-PEG1-N3 is an E3 ligase ligand-linker conjugate designed for use in PROTAC development and bioorthogonal labeling. It couples an AHPC-derived VHL ligand to a PEG1 spacer terminated with an azide for conjugation chemistry.

  • Synthesized E3 ligase ligand-linker conjugate incorporating an AHPC VHL ligand.
  • Peg1 spacer with a terminal azide enabling click chemistry conjugation.
  • Molecular weight 557.67 g/mol and formula C26H35N7O5S.
  • High purity (98.78%) suitable for chemical biology applications.
  • Soluble in DMSO (100 mg/mL) and essentially insoluble in water.
  • Supplied as a powder with recommended storage at -20°C for long-term stability.

Catalog No. 50-002-44952 Supplier Medchemexpress LLC Supplier No. HY1036001G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG1-N3 is an E3 ligase ligand-linker conjugate designed for use in PROTAC development and bioorthogonal labeling. It couples an AHPC-derived VHL ligand to a PEG1 spacer terminated with an azide for conjugation chemistry.

  • Synthesized E3 ligase ligand-linker conjugate incorporating an AHPC VHL ligand.
  • Peg1 spacer with a terminal azide enabling click chemistry conjugation.
  • Molecular weight 557.67 g/mol and formula C26H35N7O5S.
  • High purity (98.78%) suitable for chemical biology applications.
  • Soluble in DMSO (100 mg/mL) and essentially insoluble in water.
  • Supplied as a powder with recommended storage at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.