Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-acid | 2172820-09-4 | 98.9% | C30H42N4O8S | 250 MG
SDP
Catalog No. 5000352879
Encompass_Preferred
(S,R,S)-AHPC-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteeosome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52879 Supplier Medchemexpress LLC Supplier No. 250MGHY141016
(S,R,S)-AHPC-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteeosome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only