Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-PEG2-C4-Cl | 1835705-57-1 | MFCD31560480 | 98.2% | 651.26 g/mol | C32H47ClN4O6S | 2 G
SDP

Catalog No. 5000247641
Encompass_Preferred

A VHL ligand-linker conjugate used as a PROTAC building block; it incorporates the (S,R,S)-AHPC VHL ligand connected via a PEG2-C4 alkyl/ether linker with a terminal chloride and has been reported to induce degradation of GFP-HaloTag7 in cell-based assays.

  • Used as a building block for PROTAC synthesis and E3 ligase ligand-linker conjugates.
  • Contains a VHL-binding (S,R,S)-AHPC motif with a PEG2-C4 linker.
  • Reported to induce GFP-HaloTag7 degradation in cell assays.
  • Soluble in DMSO, water, and ethanol at approximately 50 mg/mL (may require ultrasonic).
  • Available in multiple package sizes, including a 2 g option.

Catalog No. 50-002-47641 Supplier Medchemexpress LLC Supplier No. HY1036072G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A VHL ligand-linker conjugate used as a PROTAC building block; it incorporates the (S,R,S)-AHPC VHL ligand connected via a PEG2-C4 alkyl/ether linker with a terminal chloride and has been reported to induce degradation of GFP-HaloTag7 in cell-based assays.

  • Used as a building block for PROTAC synthesis and E3 ligase ligand-linker conjugates.
  • Contains a VHL-binding (S,R,S)-AHPC motif with a PEG2-C4 linker.
  • Reported to induce GFP-HaloTag7 degradation in cell assays.
  • Soluble in DMSO, water, and ethanol at approximately 50 mg/mL (may require ultrasonic).
  • Available in multiple package sizes, including a 2 g option.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.