Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-N3 | 2010159-45-0 | 99.9% | 601.72 g/mol | C28H39N7O6S | 5 MG
SDP
Catalog No. 5000245148
Encompass_Preferred
(S,R,S)-AHPC-PEG2-N3 is an E3 ligase ligand-linker conjugate bearing a terminal azide for click chemistry, used as a building block in PROTAC synthesis and bioconjugation.
  • Terminal azide enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Two-unit PEG linker provides solubility and spacer functionality.
  • Designed for conjugation to alkyne, DBCO, or BCN functional partners.
  • Supplied in small-quantity formats for research and synthesis workflows.
  • Molecular weight 601.72 g/mol; molecular formula C28H39N7O6S.
Catalog No. 50-002-45148 Supplier Medchemexpress LLC Supplier No. HY1035995MG
(S,R,S)-AHPC-PEG2-N3 is an E3 ligase ligand-linker conjugate bearing a terminal azide for click chemistry, used as a building block in PROTAC synthesis and bioconjugation.
  • Terminal azide enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Two-unit PEG linker provides solubility and spacer functionality.
  • Designed for conjugation to alkyne, DBCO, or BCN functional partners.
  • Supplied in small-quantity formats for research and synthesis workflows.
  • Molecular weight 601.72 g/mol; molecular formula C28H39N7O6S.