Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-NH2 | 2010159-60-9 | 95.4% | 575.72 g·mol⁻¹ | C28H41N5O6S | 50 MG
SDP
Catalog No. 5000244953
Encompass_Preferred
(S,R,S)-AHPC-PEG2-NH2 is an E3 ligase VHL ligand-linker conjugate that links an (S,R,S)-configured AHPC VHL ligand to a two-unit polyethylene glycol (PEG2) spacer ending in a primary amine, used as a building block in PROTAC synthesis.
  • VHL ligand-linker conjugate for targeted protein degradation research.
  • Contains an (S,R,S)-configured AHPC VHL ligand.
  • Features a two-unit PEG linker terminating in a primary amine for conjugation.
  • Provided as a free base solid, typically white to yellow.
  • Available in small research pack sizes suitable for synthesis workflows.
  • High reported purity appropriate for medicinal chemistry and probe synthesis.
Catalog No. 50-002-44953 Supplier Medchemexpress LLC Supplier No. HY103603A50MG
(S,R,S)-AHPC-PEG2-NH2 is an E3 ligase VHL ligand-linker conjugate that links an (S,R,S)-configured AHPC VHL ligand to a two-unit polyethylene glycol (PEG2) spacer ending in a primary amine, used as a building block in PROTAC synthesis.
  • VHL ligand-linker conjugate for targeted protein degradation research.
  • Contains an (S,R,S)-configured AHPC VHL ligand.
  • Features a two-unit PEG linker terminating in a primary amine for conjugation.
  • Provided as a free base solid, typically white to yellow.
  • Available in small research pack sizes suitable for synthesis workflows.
  • High reported purity appropriate for medicinal chemistry and probe synthesis.