Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-NH2 dihydrochloride | 2341796-76-5 | 648.64 g/mol | C28H43Cl2N5O6S | 100 MG
SDP
Catalog No. 5000247675
Encompass_Preferred
(S,R,S)-AHPC-PEG2-NH2 dihydrochloride is a VHL E3 ligase ligand-linker conjugate used as a building block in PROTAC synthesis. It combines an (S,R,S)-configured AHPC VHL ligand with a two-unit PEG linker terminating in a primary amine and is supplied as a dihydrochloride salt in powder form for research use.
  • VHL ligand-linker conjugate for PROTAC assembly.
  • Two-unit PEG linker with terminal primary amine.
  • Dihydrochloride salt stable as a sealed powder.
  • Supplied in small milligram quantities for medicinal chemistry workflows.
  • Suitable as a synthetic building block in degrader design.
Catalog No. 50-002-47675 Supplier Medchemexpress LLC Supplier No. HY103603B100MG
(S,R,S)-AHPC-PEG2-NH2 dihydrochloride is a VHL E3 ligase ligand-linker conjugate used as a building block in PROTAC synthesis. It combines an (S,R,S)-configured AHPC VHL ligand with a two-unit PEG linker terminating in a primary amine and is supplied as a dihydrochloride salt in powder form for research use.
  • VHL ligand-linker conjugate for PROTAC assembly.
  • Two-unit PEG linker with terminal primary amine.
  • Dihydrochloride salt stable as a sealed powder.
  • Supplied in small milligram quantities for medicinal chemistry workflows.
  • Suitable as a synthetic building block in degrader design.