Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-ahpc-peg3-n3 | 1797406-80-4 | 99.7% | 645.77 g·mol⁻¹ | C30H43N7O7S | 25 MG
SDP

Catalog No. 5000247636
Encompass_Preferred

(S,R,S)-AHPC-PEG3-N3 is a VHL E3 ligase ligand-linker conjugate containing a three-unit polyethylene glycol (PEG3) spacer terminated with an azide group. It is designed as a PROTAC building block and for click chemistry conjugation to alkyne-bearing molecules.

  • VHL ligand for recruiting von Hippel-Lindau E3 ligase.
  • PEG3 spacer with a terminal azide suitable for copper-catalyzed azide-alkyne cycloaddition.
  • Intended for PROTAC synthesis and small-molecule conjugation workflows.
  • Molecular weight 645.77 g·mol⁻¹; formula C30H43N7O7S.
  • High purity (>99.6% by LCMS), suitable for research applications.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-47636 Supplier Medchemexpress LLC Supplier No. HY10359825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG3-N3 is a VHL E3 ligase ligand-linker conjugate containing a three-unit polyethylene glycol (PEG3) spacer terminated with an azide group. It is designed as a PROTAC building block and for click chemistry conjugation to alkyne-bearing molecules.

  • VHL ligand for recruiting von Hippel-Lindau E3 ligase.
  • PEG3 spacer with a terminal azide suitable for copper-catalyzed azide-alkyne cycloaddition.
  • Intended for PROTAC synthesis and small-molecule conjugation workflows.
  • Molecular weight 645.77 g·mol⁻¹; formula C30H43N7O7S.
  • High purity (>99.6% by LCMS), suitable for research applications.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.