Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-N3 | 1797406-80-4 | MFCD31807299 | >95.0% | 645.77 g/mol | C30H43N7O7S | 500 MG
SDP
Catalog No. 5000245011
Encompass_Preferred
(S,R,S)-AHPC-PEG3-N3 is a VHL (E3 ligase) ligand-linker conjugate bearing a terminal azide (-N3) for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used as a building block for PROTACs and bifunctional probes, enabling VHL recruitment and modular conjugation via click chemistry.
  • E3 ligase (VHL) ligand-linker conjugate suitable for PROTAC design.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Three-unit PEG linker provides flexibility and spacing between ligand and cargo.
  • Useful as a building block for bifunctional molecules and bioconjugation.
  • Reported appearance: solid-liquid mixture; CAS 1797406-80-4; molecular weight 645.77 g/mol.
Catalog No. 50-002-45011 Supplier Medchemexpress LLC Supplier No. HY103598500MG
(S,R,S)-AHPC-PEG3-N3 is a VHL (E3 ligase) ligand-linker conjugate bearing a terminal azide (-N3) for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used as a building block for PROTACs and bifunctional probes, enabling VHL recruitment and modular conjugation via click chemistry.
  • E3 ligase (VHL) ligand-linker conjugate suitable for PROTAC design.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Three-unit PEG linker provides flexibility and spacing between ligand and cargo.
  • Useful as a building block for bifunctional molecules and bioconjugation.
  • Reported appearance: solid-liquid mixture; CAS 1797406-80-4; molecular weight 645.77 g/mol.