Learn More
Medchemexpress LLC HY-103602A 100mg Medchemexpress, (S,R,S)-AHPC-PEG3-NH2 CAS:2010159-56-3 Purity:>98%
SDP
Catalog No. 501968137
Encompass_Preferred
Medchemexpress, HY-103602A 100mg (S,R,S)-AHPC-PEG3-NH2 CAS:2010159-56-3 (S,R,S)-AHPC-PEG3-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8137 Supplier Medchemexpress LLC Supplier No. HY103602A100MG
Medchemexpress, HY-103602A 100mg (S,R,S)-AHPC-PEG3-NH2 CAS:2010159-56-3 (S,R,S)-AHPC-PEG3-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.