Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 | 2010159-56-3 | MFCD31727698 | 99.3% | 619.80 g/mol | C30H45N5O7S | 500 MG
SDP
Catalog No. 5000244593
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 is a VHL E3 ligase ligand-linker conjugate terminating in a primary amine. It is supplied as a research-grade powder for use as a building block in PROTAC synthesis and related chemical biology applications.
  • VHL-binding motif for targeted protein degradation studies.
  • Three-unit PEG linker with terminal amine for facile conjugation.
  • High purity suitable for synthetic applications.
  • Supplied as a powder convenient for storage and handling.
  • Store and handle per the manufacturer's datasheet recommendations.
Catalog No. 50-002-44593 Supplier Medchemexpress LLC Supplier No. HY103602A500MG
(S,R,S)-AHPC-PEG3-NH2 is a VHL E3 ligase ligand-linker conjugate terminating in a primary amine. It is supplied as a research-grade powder for use as a building block in PROTAC synthesis and related chemical biology applications.
  • VHL-binding motif for targeted protein degradation studies.
  • Three-unit PEG linker with terminal amine for facile conjugation.
  • High purity suitable for synthetic applications.
  • Supplied as a powder convenient for storage and handling.
  • Store and handle per the manufacturer's datasheet recommendations.