Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | >95.0% | 656.23 | C30H46ClN5O7S | 2 G
SDP

Catalog No. 5000247626
Encompass_Preferred

(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for use in PROTAC development and related chemical biology research. It is a PEG3-linked derivative of the AHPC VHL ligand and is provided in multiple packaged sizes for synthesis and assay workflows.

  • VHL E3 ligase ligand-linker conjugate suitable for PROTAC design.
  • Provided as the hydrochloride salt for improved solubility and handling.
  • Available in multiple sizes, including 2 G.
  • Molecular formula C30H46ClN5O7S; molecular weight 656.23.
  • Purity ≥95% by supplier specification.

Catalog No. 50-002-47626 Supplier Medchemexpress LLC Supplier No. HY1036022G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for use in PROTAC development and related chemical biology research. It is a PEG3-linked derivative of the AHPC VHL ligand and is provided in multiple packaged sizes for synthesis and assay workflows.

  • VHL E3 ligase ligand-linker conjugate suitable for PROTAC design.
  • Provided as the hydrochloride salt for improved solubility and handling.
  • Available in multiple sizes, including 2 G.
  • Molecular formula C30H46ClN5O7S; molecular weight 656.23.
  • Purity ≥95% by supplier specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.