Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | 99.0% | 656.23 g/mol | C30H46ClN5O7S | 1 G
SDP
Catalog No. 5000245157
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate that combines a stereodefined AHPC VHL ligand with a three-unit polyethylene glycol linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a research building block for PROTAC synthesis and other chemical biology applications.
  • Hydrochloride salt form for improved handling.
  • Primary amine terminus suitable for common coupling chemistries.
  • Three-unit PEG linker provides aqueous solubility and flexible spacing.
  • Stereochemically defined (S,R,S) VHL ligand for targeted E3 ligase engagement.
  • High reported purity (~99.0%) for reliable synthetic use.
  • Available in small to gram-scale quantities for research workflows.
Catalog No. 50-002-45157 Supplier Medchemexpress LLC Supplier No. HY1036021G
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate that combines a stereodefined AHPC VHL ligand with a three-unit polyethylene glycol linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a research building block for PROTAC synthesis and other chemical biology applications.
  • Hydrochloride salt form for improved handling.
  • Primary amine terminus suitable for common coupling chemistries.
  • Three-unit PEG linker provides aqueous solubility and flexible spacing.
  • Stereochemically defined (S,R,S) VHL ligand for targeted E3 ligase engagement.
  • High reported purity (~99.0%) for reliable synthetic use.
  • Available in small to gram-scale quantities for research workflows.