Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | 99.0% | 656.23 g/mol | C30H46ClN5O7S | 10 MG
SDP
Catalog No. 5000245149
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a three-unit PEG linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a building block in PROTAC synthesis to recruit the VHL E3 ligase.
  • hydrochloride salt form for improved handling and stability.
  • three-unit PEG linker terminating in a primary amine for conjugation.
  • suitable as a VHL E3 ligase recruiter in PROTAC design.
  • high purity (99.0%) for research applications.
  • small pack sizes available (1 mg, 5 mg, 10 mg) for early-stage studies.
Catalog No. 50-002-45149 Supplier Medchemexpress LLC Supplier No. HY10360210MG
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a three-unit PEG linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a building block in PROTAC synthesis to recruit the VHL E3 ligase.
  • hydrochloride salt form for improved handling and stability.
  • three-unit PEG linker terminating in a primary amine for conjugation.
  • suitable as a VHL E3 ligase recruiter in PROTAC design.
  • high purity (99.0%) for research applications.
  • small pack sizes available (1 mg, 5 mg, 10 mg) for early-stage studies.