Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | 99.0% | 656.23 g/mol | C30H46ClN5O7S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY103602100MG
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate functionalized with a PEG3 amine, provided as the hydrochloride salt and intended for use as a building block in PROTAC and targeted protein degradation research.
  • Hydrochloride salt supplied as a solid, light yellow to yellow.
  • High reported purity (≈99%).
  • Molecular weight ~656.23 g/mol (hydrochloride form).
  • PEG3 amine functionalization enables linker conjugation to target ligands.
  • Intended for use in medicinal chemistry and chemical biology applications.
Catalog No. 50-002-47791