Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | 99.0% | 656.23 g/mol | C30H46ClN5O7S | 50 MG
SDP
Catalog No. 5000244609
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate featuring an AHPC-derived VHL-binding warhead connected to a three-unit polyethylene glycol spacer terminating in a primary amine; it is supplied as the hydrochloride salt and is used as a building block in PROTAC synthesis and E3 ligase ligand studies.
  • Contains AHPC-derived VHL-binding warhead with defined stereochemistry.
  • Three-unit PEG spacer terminating in a primary amine for conjugation.
  • Provided as the hydrochloride salt for improved stability and handling.
  • Intended for use in PROTAC design and E3 ligase ligand research.
  • Requires cold, inert storage and protection from light.
Catalog No. 50-002-44609 Supplier Medchemexpress LLC Supplier No. HY10360250MG
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is a VHL E3 ligase ligand-linker conjugate featuring an AHPC-derived VHL-binding warhead connected to a three-unit polyethylene glycol spacer terminating in a primary amine; it is supplied as the hydrochloride salt and is used as a building block in PROTAC synthesis and E3 ligase ligand studies.
  • Contains AHPC-derived VHL-binding warhead with defined stereochemistry.
  • Three-unit PEG spacer terminating in a primary amine for conjugation.
  • Provided as the hydrochloride salt for improved stability and handling.
  • Intended for use in PROTAC design and E3 ligase ligand research.
  • Requires cold, inert storage and protection from light.