Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 hydrochloride | 2097971-11-2 | MFCD31656714 | 99.0% | 656.23 g/mol | C30H46ClN5O7S | 500 MG
SDP
Catalog No. 5000245016
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC-based VHL ligand with a three-unit polyethylene glycol (PEG3) linker terminating in a primary amine. It is supplied as the hydrochloride salt and is used as a building block in PROTAC synthesis and other chemical biology applications.
  • E3 ligase VHL ligand-linker conjugate for targeted protein degradation research.
  • PEG3 linker with terminal primary amine suitable for conjugation.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (≈99%) suitable for synthetic applications.
  • Available in multiple preweighed sizes including 500 MG.
Catalog No. 50-002-45016 Supplier Medchemexpress LLC Supplier No. HY103602500MG
(S,R,S)-AHPC-PEG3-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC-based VHL ligand with a three-unit polyethylene glycol (PEG3) linker terminating in a primary amine. It is supplied as the hydrochloride salt and is used as a building block in PROTAC synthesis and other chemical biology applications.
  • E3 ligase VHL ligand-linker conjugate for targeted protein degradation research.
  • PEG3 linker with terminal primary amine suitable for conjugation.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (≈99%) suitable for synthetic applications.
  • Available in multiple preweighed sizes including 500 MG.