Learn More
TARGETMOL CHEMICALS INC S R S-AHPC-PEG4-N3 25MG
SDP
Catalog No. 502878614
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (S,R,S)-AHPC-PEG4-N3 (VHL Ligand-Linker Conjugates 5) is a synthesized ligand-linker for E3 ligase-based systems, comprising the (S,R,S)-AHPC VHL ligand and a 4-unit PEG linker, commonly used in PROTAC technology. Purity 98.06%
Catalog No. 50-287-8614 Supplier TARGETMOL CHEMICALS INC Supplier No. T1792125MG
Also available in 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (S,R,S)-AHPC-PEG4-N3 (VHL Ligand-Linker Conjugates 5) is a synthesized ligand-linker for E3 ligase-based systems, comprising the (S,R,S)-AHPC VHL ligand and a 4-unit PEG linker, commonly used in PROTAC technology. Purity 98.06%