Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-PEG4-NH2 | 2010159-57-4 | MFCD31657338 | 95.6% | 663.83 | C32H49N5O8S | 25 MG
SDP

Catalog No. 5000245155
Encompass_Preferred

(S,R,S)-AHPC-PEG4-NH2 is an E3 ligase ligand-linker conjugate that combines a VHL-targeting AHPC ligand with a four-unit PEG linker for use in PROTAC design and targeted protein degradation research. It is supplied as a solid powder for laboratory synthesis and biochemical studies.

  • Designed for VHL-mediated targeted protein degradation applications.
  • Contains a four-unit polyethylene glycol linker to provide flexibility and solubility.
  • White to off-white solid powder form suitable for synthesis workflows.
  • CAS number 2010159-57-4 for unambiguous identification.
  • Molecular weight 663.83 g·mol-1 and formula C32H49N5O8S.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-45155 Supplier Medchemexpress LLC Supplier No. HY103604A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG4-NH2 is an E3 ligase ligand-linker conjugate that combines a VHL-targeting AHPC ligand with a four-unit PEG linker for use in PROTAC design and targeted protein degradation research. It is supplied as a solid powder for laboratory synthesis and biochemical studies.

  • Designed for VHL-mediated targeted protein degradation applications.
  • Contains a four-unit polyethylene glycol linker to provide flexibility and solubility.
  • White to off-white solid powder form suitable for synthesis workflows.
  • CAS number 2010159-57-4 for unambiguous identification.
  • Molecular weight 663.83 g·mol-1 and formula C32H49N5O8S.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.