Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG4-NH2 (hydrochloride) | 2010159-57-4 | MFCD31656715 | ≥95.0% | 663.83 g/mol | C32H49N5O8S | 100 MG
SDP
Catalog No. 5000244991
Encompass_Preferred
(S,R,S)-AHPC-PEG4-NH2 is a VHL E3 ligase ligand-linker conjugate designed for use as a building block in PROTAC synthesis. It combines an (S,R,S)-AHPC-derived VHL ligand with a four-unit polyethylene glycol spacer terminating in a primary amine to provide a conjugation handle for bifunctional degrader assembly.
  • VHL ligand with PEG4 spacer and terminal primary amine.
  • Provides a conjugation handle for PROTAC assembly.
  • Solid form suitable for small-scale synthesis and derivatization.
  • Compatible with common organic solvents for coupling reactions.
  • Store sealed at recommended temperatures; follow SDS for handling and safety.
Catalog No. 50-002-44991 Supplier Medchemexpress LLC Supplier No. HY103604A100MG
(S,R,S)-AHPC-PEG4-NH2 is a VHL E3 ligase ligand-linker conjugate designed for use as a building block in PROTAC synthesis. It combines an (S,R,S)-AHPC-derived VHL ligand with a four-unit polyethylene glycol spacer terminating in a primary amine to provide a conjugation handle for bifunctional degrader assembly.
  • VHL ligand with PEG4 spacer and terminal primary amine.
  • Provides a conjugation handle for PROTAC assembly.
  • Solid form suitable for small-scale synthesis and derivatization.
  • Compatible with common organic solvents for coupling reactions.
  • Store sealed at recommended temperatures; follow SDS for handling and safety.