Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG5-COOH | 2172820-14-1 | 99.8% | 1 ML
SDP
Catalog No. 5000338634
Encompass_Preferred
(S,R,S)-AHPC-PEG5-COOH (VH032-PEG5-COOH) is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 5-unit PEG linker used in PROTAC technology. This product is for research use only and not sold to patients.
  • Incorporates an (S,R,S)-AHPC-based VHL ligand
  • Features a 5-unit PEG linker
  • Utilized in PROTAC technology
  • Recruits VHL protein, a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes
  • Induces ubiquitination and subsequent proteasomal degradation of target proteins
Catalog No. 50-003-38634 Supplier Medchemexpress LLC Supplier No. HY13027110MM/1ML
(S,R,S)-AHPC-PEG5-COOH (VH032-PEG5-COOH) is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 5-unit PEG linker used in PROTAC technology. This product is for research use only and not sold to patients.
  • Incorporates an (S,R,S)-AHPC-based VHL ligand
  • Features a 5-unit PEG linker
  • Utilized in PROTAC technology
  • Recruits VHL protein, a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes
  • Induces ubiquitination and subsequent proteasomal degradation of target proteins