Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG5-COOH | 2172820-14-1 | 99.8% | 25 MG
SDP
Catalog No. 5000342312
Encompass_Preferred
(S,R,S)-AHPC-PEG5-COOH is a synthesized E3 ligase ligand-linker conjugate designed for PROTAC technology. It incorporates an (S,R,S)-AHPC based VHL ligand and a 5-unit PEG linker. This compound targets the VHL protein, a key subunit of Cullin RING E3 ubiquitin ligase complexes, facilitating ubiquitination and subsequent proteasomal degradation of target proteins. It is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Features a 5-unit PEG linker
  • Utilized in PROTAC technology
  • Recruits VHL protein to induce target protein degradation
  • For research use only
  • Appears as a colorless to off-white solid
  • Store at -20°C under nitrogen
Catalog No. 50-003-42312 Supplier Medchemexpress LLC Supplier No. 25MGHY130271
(S,R,S)-AHPC-PEG5-COOH is a synthesized E3 ligase ligand-linker conjugate designed for PROTAC technology. It incorporates an (S,R,S)-AHPC based VHL ligand and a 5-unit PEG linker. This compound targets the VHL protein, a key subunit of Cullin RING E3 ubiquitin ligase complexes, facilitating ubiquitination and subsequent proteasomal degradation of target proteins. It is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Features a 5-unit PEG linker
  • Utilized in PROTAC technology
  • Recruits VHL protein to induce target protein degradation
  • For research use only
  • Appears as a colorless to off-white solid
  • Store at -20°C under nitrogen