Learn More
Medchemexpress LLC HY-103606 100mg Medchemexpress, (S,R,S)-AHPC-PEG6-C4-Cl CAS:1835705-59-3 Purity:>98%
SDP
Catalog No. 501968141
Encompass_Preferred
Medchemexpress, HY-103606 100mg (S,R,S)-AHPC-PEG6-C4-Cl CAS:1835705-59-3 (S,R,S)-AHPC-PEG6-C4-Cl is a small molecule HaloPROTAC that incorporates the (S,R,S)-AHPC based VHL ligand and 6-unit PEG linker. (S,R,S)-AHPC-PEG6-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8141 Supplier Medchemexpress LLC Supplier No. HY103606100MG
Medchemexpress, HY-103606 100mg (S,R,S)-AHPC-PEG6-C4-Cl CAS:1835705-59-3 (S,R,S)-AHPC-PEG6-C4-Cl is a small molecule HaloPROTAC that incorporates the (S,R,S)-AHPC based VHL ligand and 6-unit PEG linker. (S,R,S)-AHPC-PEG6-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.