Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride | 2376990-28-0 | 98.2% | C30H47Cl2N5O5S | 10MG
SDP
Catalog No. 5000261787
Encompass_Preferred
(S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride is a VH032-derived VHL E3 ligase ligand-linker conjugate provided as the dihydrochloride salt. It serves as a building block for PROTAC degraders and related chemical biology reagents, offering a functionalized phenol-alkyl spacer for conjugation to target ligands.
  • High purity (98.2%) suitable for synthetic and biological applications.
  • Available as a solid with light yellow to yellow appearance.
  • Includes a functionalized phenol-alkyl C6 amine linker for conjugation.
  • Molecular formula C30H47Cl2N5O5S; molecular weight 660.70 g·mol⁻¹.
  • Intended for research use; follow proper handling and storage instructions.
Catalog No. 50-002-61787 Supplier Medchemexpress LLC Supplier No. HY13618310MG
(S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride is a VH032-derived VHL E3 ligase ligand-linker conjugate provided as the dihydrochloride salt. It serves as a building block for PROTAC degraders and related chemical biology reagents, offering a functionalized phenol-alkyl spacer for conjugation to target ligands.
  • High purity (98.2%) suitable for synthetic and biological applications.
  • Available as a solid with light yellow to yellow appearance.
  • Includes a functionalized phenol-alkyl C6 amine linker for conjugation.
  • Molecular formula C30H47Cl2N5O5S; molecular weight 660.70 g·mol⁻¹.
  • Intended for research use; follow proper handling and storage instructions.