Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride | 2376990-26-8 | 99.0% | 632.64 g/mol | C28H43Cl2N5O5S | 5 MG
SDP
Catalog No. 5000222106
Encompass_Preferred
(S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride is a von Hippel-Lindau (VHL) ligand building block used in PROTAC and E3 ligase chemistry. Supplied as a high-purity, research-scale solid, it is suitable for synthetic chemistry and ligand design workflows.
  • VHL ligand used to recruit the von Hippel-Lindau protein.
  • High reported purity (99.01%).
  • White to off-white solid physical form.
  • Available in small research-scale quantities, including 5 MG.
  • Datasheet, certificate of analysis, and safety data sheet available for characterization and handling.
Catalog No. 50-002-22106 Supplier Medchemexpress LLC Supplier No. HY1361845MG
(S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride is a von Hippel-Lindau (VHL) ligand building block used in PROTAC and E3 ligase chemistry. Supplied as a high-purity, research-scale solid, it is suitable for synthetic chemistry and ligand design workflows.
  • VHL ligand used to recruit the von Hippel-Lindau protein.
  • High reported purity (99.01%).
  • White to off-white solid physical form.
  • Available in small research-scale quantities, including 5 MG.
  • Datasheet, certificate of analysis, and safety data sheet available for characterization and handling.