Learn More
Abcam (S)-(-)-Raclopride, D2 receptor antagonist, 50MG
Catalog No. 502707482
Encompass_Preferred
MW 347.24 g/mol, Purity >97%. High affinity (Kd = 1.2 nM) and high selectivity for D₂ receptors in vitro and in vivo (IC₅₀ = 32 nM). Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7482 Supplier Abcam Supplier No. AB12056350MG
MW 347.24 g/mol, Purity >97%. High affinity (Kd = 1.2 nM) and high selectivity for D₂ receptors in vitro and in vivo (IC₅₀ = 32 nM). Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement