Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-remoxipride hydrochloride | 73220-03-8 | 99.9% | 371.27 g/mol | C16H23BrN2O3 | 10 MG
SDP

Catalog No. 5000211294
Encompass_Preferred

(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride supplied as the hydrochloride salt for laboratory research. It is a selective dopamine D2 receptor antagonist used in neuroscience and pharmacology studies, supplied as a solid powder with reported high purity and characterized by CAS 73220-03-8.

  • Selective dopamine D2 receptor antagonist for receptor pharmacology studies.
  • S-enantiomeric form for stereospecific research.
  • Hydrochloride salt form, solid powder suitable for in vitro use.
  • High purity, typically reported as 99.9%.
  • Molecular weight 371.27 g/mol and formula C16H23BrN2O3.
  • Intended for research use only; not for human or clinical applications.

Catalog No. 50-002-11294 Supplier Medchemexpress LLC Supplier No. HY101313A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride supplied as the hydrochloride salt for laboratory research. It is a selective dopamine D2 receptor antagonist used in neuroscience and pharmacology studies, supplied as a solid powder with reported high purity and characterized by CAS 73220-03-8.

  • Selective dopamine D2 receptor antagonist for receptor pharmacology studies.
  • S-enantiomeric form for stereospecific research.
  • Hydrochloride salt form, solid powder suitable for in vitro use.
  • High purity, typically reported as 99.9%.
  • Molecular weight 371.27 g/mol and formula C16H23BrN2O3.
  • Intended for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.