Learn More
Medchemexpress LLC (S)-remoxipride hydrochloride | 73220-03-8 | 99.9% | 407.73 g/mol | C16H24BrClN2O3 | 100 MG
SDP
Catalog No. 5000211291
Encompass_Preferred
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride, provided as a research-grade dopamine D2 receptor antagonist for preclinical studies of psychotic disorders and dopaminergic signaling. Handle according to the supplier safety data sheet and use analytical methods to confirm batch-specific purity and form.
  • Selective dopamine D2 receptor antagonist.
  • Used in research on psychotic disorders and dopaminergic pathways.
  • High reported purity suitable for analytical and biological studies.
  • Supplied as small milligram quantities for laboratory use.
Catalog No. 50-002-11291 Supplier Medchemexpress LLC Supplier No. HY101313A100MG
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride, provided as a research-grade dopamine D2 receptor antagonist for preclinical studies of psychotic disorders and dopaminergic signaling. Handle according to the supplier safety data sheet and use analytical methods to confirm batch-specific purity and form.
  • Selective dopamine D2 receptor antagonist.
  • Used in research on psychotic disorders and dopaminergic pathways.
  • High reported purity suitable for analytical and biological studies.
  • Supplied as small milligram quantities for laboratory use.