Learn More
Medchemexpress LLC (S)-Remoxipride hydrochloride | 73220-03-8 | 99.9% | 407.8 g·mol⁻¹ | C16H23BrN2O3·HCl | 50 MG
SDP
Catalog No. 5000211380
Encompass_Preferred
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride, supplied as the hydrochloride salt for research use. It acts as a selective dopamine D2 receptor antagonist and is provided at high purity for in vitro and in vivo pharmacology studies.
  • S-enantiomer with selective dopamine D2 receptor antagonist activity.
  • Provided as the hydrochloride salt, white to off-white solid.
  • High purity suitable for research applications.
  • Commonly used in receptor binding and pharmacological assays.
  • Available in small research quantities (e.g., 50 mg).
Catalog No. 50-002-11380 Supplier Medchemexpress LLC Supplier No. HY101313A50MG
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride, supplied as the hydrochloride salt for research use. It acts as a selective dopamine D2 receptor antagonist and is provided at high purity for in vitro and in vivo pharmacology studies.
  • S-enantiomer with selective dopamine D2 receptor antagonist activity.
  • Provided as the hydrochloride salt, white to off-white solid.
  • High purity suitable for research applications.
  • Commonly used in receptor binding and pharmacological assays.
  • Available in small research quantities (e.g., 50 mg).