Learn More
Medchemexpress LLC (S)-Remoxipride hydrochloride | 73220-03-8 | MFCD01743944 | >98.0% | 407.7 g/mol | C16H23BrN2O3.HCl | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101313A25MG
Encompass_Preferred
(S)-Remoxipride hydrochloride is the S-enantiomer salt of remoxipride, a selective dopamine D2 receptor antagonist used as a research reagent in receptor pharmacology and biochemical studies. The hydrochloride salt improves aqueous solubility and the material is typically provided in small, high-purity milligram quantities.
  • Selective dopamine D2 receptor antagonist for receptor pharmacology studies.
  • Enantiomerically defined S-form for stereoselective research.
  • High purity suitable for biochemical and cellular assays.
  • Hydrochloride salt form enhances solubility in aqueous media.
  • Available in small milligram pack sizes for laboratory workflows.
Catalog No. 50-002-11216