Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sörensen's Sodium Phosphate Buffer, 0.1 M Total Phosphate, pH 7.15, Ricca Chemical
SDP

Catalog No. 79885 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7988-5 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7988-5 Supplier Ricca Chemical Company Supplier No. 79885
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Sörensen's Sodium Phosphate Buffer
CAS Min % 97.0, 0.99, 0.38, 0.01
CAS Max % 100.0, 1.03, 0.39, 0.01
CAS 7558-79-4, 10049-21-5, 89-83-8
Concentration 0.1 M
For Use With (Application) Water Analysis
For Use With (Equipment) Ion Selective Electrode
pH 7.15
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.