Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-rivastigmine-d4 | 123441-03-2 | MFCD00871496 | >98.0% | 254.36 g/mol | C14H18D4N2O2 | 1 MG
SDP

Catalog No. 5000284034
Encompass_Preferred

(S)-Rivastigmine-d4 is a deuterium-labeled isotopologue of rivastigmine supplied in milligram quantities as an analytical standard for research and quantitative mass spectrometry. It is intended for use as an isotope-labeled internal standard or tracer in studies of cholinesterase activity, pharmacokinetics, and metabolic profiling.

  • Deuterium-labeled internal standard (d4)
  • Chemical formula: C14H18D4N2O2
  • Molecular weight: 254.36 g/mol
  • Purity: >98.0%
  • Suitable for LC-MS/MS quantitation and metabolic studies
  • Provided as a 1 mg vial for analytical use

Catalog No. 50-002-84034 Supplier Medchemexpress LLC Supplier No. HYW6543361MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Rivastigmine-d4 is a deuterium-labeled isotopologue of rivastigmine supplied in milligram quantities as an analytical standard for research and quantitative mass spectrometry. It is intended for use as an isotope-labeled internal standard or tracer in studies of cholinesterase activity, pharmacokinetics, and metabolic profiling.

  • Deuterium-labeled internal standard (d4)
  • Chemical formula: C14H18D4N2O2
  • Molecular weight: 254.36 g/mol
  • Purity: >98.0%
  • Suitable for LC-MS/MS quantitation and metabolic studies
  • Provided as a 1 mg vial for analytical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.