Learn More
Medchemexpress LLC (S)-rivastigmine-d6 tartrate | 194930-00-2 | 99.0% | 406.46 | C18H22D6N2O8 | 5 MG
SDP
Catalog No. 5000263548
Encompass_Preferred
(S)-Rivastigmine-d6 tartrate is the deuterium-labeled form of (S)-rivastigmine tartrate supplied as a solid research standard for use as an analytical tracer, internal standard, or in quantitative studies of cholinesterase inhibition. It is intended for laboratory research and includes recommended storage and solvent guidelines for reliable handling.
  • Deuterium-labeled internal standard for quantitative analysis
  • High isotopic and chemical purity (99.0%)
  • Stable solid with recommended storage at -20°C; in solution store at -80°C for long-term stability
  • Soluble in DMF, DMSO, and ethanol at specified concentrations
  • Available in small milligram quantities for analytical workflows
Catalog No. 50-002-63548 Supplier Medchemexpress LLC Supplier No. HY11017AS5MG
(S)-Rivastigmine-d6 tartrate is the deuterium-labeled form of (S)-rivastigmine tartrate supplied as a solid research standard for use as an analytical tracer, internal standard, or in quantitative studies of cholinesterase inhibition. It is intended for laboratory research and includes recommended storage and solvent guidelines for reliable handling.
  • Deuterium-labeled internal standard for quantitative analysis
  • High isotopic and chemical purity (99.0%)
  • Stable solid with recommended storage at -20°C; in solution store at -80°C for long-term stability
  • Soluble in DMF, DMSO, and ethanol at specified concentrations
  • Available in small milligram quantities for analytical workflows