Learn More
Abcam (S)-(+)-Rolipram, PDE4 inhibitor, 10MG
Catalog No. 502707916
Encompass_Preferred
MW 275.34 Da, Purity >99%. PDE4 inhibitor (IC₅₀ = 0.58 μM). Less active enantiomer. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7916 Supplier Abcam Supplier No. AB12003010MG
Add to Cart
Add to Cart
MW 275.34 Da, Purity >99%. PDE4 inhibitor (IC₅₀ = 0.58 μM). Less active enantiomer. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement