Learn More
Enzo Life Sciences (S)-Roscovitine (1mg). CAS: 186692-45-5
SDP
Supplier:  Enzo Life Sciences ALX350293M001
Encompass_Preferred
(S)-Enantiomer of the CDK inhibitor roscovitine (BML-CC205). Potent inhibitor of CDK1/cyclin B kinase (IC50=950nM). Alternative name: 6-Benzylamino 2-(S)-[(1-ethyl)-2-hydroxyethylamino]-9-isopropylpurine. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO, dimethyl formamide, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8831