Learn More
AdipoGen (S)-Roscovitine (25 mg)
SDP
Supplier:  AdipoGen AGMRC0002M025
Encompass_Preferred
Chemical. CAS: 186692-45-5. Formula: C19H26N6O. MW: 354.5. (S)-enantiomer of (R)-roscovitine (Prod. No. MR-C0001). Potent selective cyclin-dependent kinase 1 (CDK1) inhibitor. Potential neuroprotectant for stroke. Crosses the blood brain barrier. Inhibits CDK5, consequently blocking hypoxia-induced apoptosis in neurons. Potential anti-inflammatory compound. Potential antidiabetic compound.
Catalog No. 50-205-4222