Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-RUCY(regR)-XylBINAP, TCI America™
SDP

Catalog No. R0138200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0138200MG 200 mg
R01381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0138200MG Supplier TCI America Supplier No. R0138200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1312713-89-5
Molecular Formula C71H73ClN2O2P2Ru
MDL Number MFCD20922903
Synonym RuCl[(S)-daipena][(S)-xylbinap]

Specifications

Color Green-Yellow
Quantity 200 mg
Formula Weight 1184.84
Physical Form Crystalline Powder
Chemical Name or Material (S)-RUCY(regR)-XylBINAP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.