Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(-)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Catalog No. AAH2598103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2598103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2598103 Supplier Thermo Scientific Chemicals Supplier No. H2598103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1052707-05-7
Molecular Formula C14H8Cl2F10N2
Molecular Weight (g/mol) 465.116
MDL Number MFCD09265323
InChI Key JXHUHXWIKZGKIE-AXEKQOJOSA-N
Synonym 1s,2s-1,2-bis 2,3,4,5,6-pentafluorophenyl ethane-1,2-diamine dihydrochloride, s,s---1,2-bis 2,3,4,5,6-pentafluorophenyl-1,2-ethanediamine dihydrochloride, s,s-1,2-bis 2,3,4,5-pentafluorophenyl-1,2-ethanediamine dihydrochloride, ee
PubChem CID 73995083
IUPAC Name (1S,2S)-1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine;dihydrochloride
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(C2=C(C(=C(C(=C2F)F)F)F)F)N)N.Cl.Cl

Specifications

Melting Point >200°C
Quantity 1 g
Optical Rotation −28° (c=1 in Methanol)
Formula Weight 465.13
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (S,S)-(-)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.