Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S″)-2,2″-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1″-biferrocene 98.0+%, TCI America™
SDP

Catalog No. B3197100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3197100MG 100 mg
B3197500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3197100MG Supplier TCI America Supplier No. B3197100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 190601-12-8
Molecular Formula C32H36Fe2N2O2
Molecular Weight (g/mol) 592.342
InChI Key GSVKKVGUOIENJR-NRMJSFJRSA-N
PubChem CID 133554196
SMILES CC(C)C1COC(=N1)[C-]2C=CC=C2C3=C(C=C[CH-]3)C4=NC(CO4)C(C)C.[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Fe+2].[Fe+2]

Specifications

Quantity 100 mg
Formula Weight 592.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S,S′′)-2,2′′-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1′′-biferrocen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.