Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine 95.0+%, TCI America™
SDP

Catalog No. B2220250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2220250MG 250 mg
B22201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2220250MG Supplier TCI America Supplier No. B2220250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 174500-20-0
Molecular Formula C23H19N3O2
Molecular Weight (g/mol) 369.424
MDL Number MFCD01863586
InChI Key HLHBIMJNCKZZQO-NHCUHLMSSA-N
Synonym 2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, 2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine, s,s-ph-pybox, 2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine, --2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, s,s, unii-5r62cxu58k, s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl, 2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
PubChem CID 7010100
IUPAC Name (4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 250 mg
Formula Weight 369.42
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.