Learn More
Chemglass Life Sciences Stainless steel adapter, M24 female to M30 male - for Huber circulator
SDP
Catalog No. NC2588386
Encompass
This stainless steel adapter converts an M24 female thread to an M30 male thread for use with Huber-style laboratory circulator connections. It provides a durable, corrosion-resistant transition fitting for matching metric circulator ports.
  • Made from stainless steel for corrosion and chemical resistance.
  • Converts M24 female threads to M30 male threads.
  • Engineered for use with Huber circulator fittings and similar laboratory circulators.
  • Sold individually for flexible replacement and installation.
Catalog No. NC2588386 Supplier Chemglass Life Sciences Supplier No. CG1969M109
This stainless steel adapter converts an M24 female thread to an M30 male thread for use with Huber-style laboratory circulator connections. It provides a durable, corrosion-resistant transition fitting for matching metric circulator ports.
  • Made from stainless steel for corrosion and chemical resistance.
  • Converts M24 female threads to M30 male threads.
  • Engineered for use with Huber circulator fittings and similar laboratory circulators.
  • Sold individually for flexible replacement and installation.