Learn More
BioSpec Products Stainless Steel Beads. 0.25 inches. 100/pack.
SDP
Supplier:  BioSpec Products 11079635SS
Encompass_Preferred
Stainless steel beads have a density of 7.9g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available. 6.35mm diameter or .25inches. 100 per pack
Catalog No. NC0251213