Learn More
Echelon Biosciences Research Labs (S,S) BISOLESYL-LBPA 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs LB181100UG
Encompass
(S,S) Bisoleoyl-lysobisphosphatidic acid is the naturally occurring, biologically activie isomer of LPBA. Lysobisphosphatidic acids (LBPAs), known also as bis-(monoacylglycerol)phosphates (BMPs) are specialized lipids reported to play a role in intracellular protein and lipid transport in healthy cells. Their accumulation was observed in pathologic liver tissue, and they also serve as antigens for auto-antibody generation in a human autoimmune condition termed antiphospholipid syndrome. Accumulation of LBPAs in intracellular, often multilamellar membranes is related to biomembrane polymorphism which may impact intracellular cholesterol transport. This item is non returnable
Catalog No. 50-720-537