Learn More
Ace Glass, Inc. S/S CLMPS SCREWLOCK SZ28 S/S CLMPS SCREWLOCK SZ28
SDP
Catalog No. 509557982
Encompass
Stainless Steel screwlock pinch clamp for use with 28/15 o-ring or ball & socket joints.
Catalog No. 50-955-7982 Supplier Ace Glass, Inc. Supplier No. 766912
Stainless Steel screwlock pinch clamp for use with 28/15 o-ring or ball & socket joints.