Learn More
AdipoGen S S-Dimethoxy-2 3-butanediol
SDP
Supplier:  AdipoGen CDXD0434G005
Encompass_Preferred
Chemical. CAS 50622-10-1. Formula C6H14O4. MW 150.17. Synthetic Building block for synthesis. For chiral auxiliary modification of LAH. Condensation of ester enolates and chiral imines to beta-lactams. Auxiliary, reactions of its acetal. Determination of the enantiomeric purity of ketones by acetal formation and 13C-NMR.
Catalog No. 50-256-1081