Learn More
Inter Dyne Systems Inc S/S DRIP TROUGH 30 L X 2 D
Catalog No. NC0404378
Encompass_Preferred
stainless steel 30 in. long x 2 in. deep drip trough with 3/4 in. deep mounting flange. Unit is supplied with a right hand location 3/ dia drain outlet and three feet of clear pvc drain tubing.
Catalog No. NC0404378 Supplier Inter Dyne Systems Inc Supplier No. HDT302
stainless steel 30 in. long x 2 in. deep drip trough with 3/4 in. deep mounting flange. Unit is supplied with a right hand location 3/ dia drain outlet and three feet of clear pvc drain tubing.