Learn More
AdipoGen S S-EDDS-Na3
SDP
Catalog No. 502560994
Encompass_Preferred
Chemical. CAS 178949-82-1. Formula C10H13N2Na3O8. MW 358.19. Synthetic Aminopolycarboxylic acid. A powerful, readily biodegradable chelant. Complexing agent, exhibiting superior biological degradation environmentally friendly compared to EDTA. Used in cosmetic formulations, as effective anti-oxidant and water hardness control, to reduce radical generation, as effective peroxide stabiliser, as preservative potentiator improving piocide efficacy and excellent transition metal chelator.
Catalog No. 50-256-0994 Supplier AdipoGen Supplier No. CDXE0016D001
Chemical. CAS 178949-82-1. Formula C10H13N2Na3O8. MW 358.19. Synthetic Aminopolycarboxylic acid. A powerful, readily biodegradable chelant. Complexing agent, exhibiting superior biological degradation environmentally friendly compared to EDTA. Used in cosmetic formulations, as effective anti-oxidant and water hardness control, to reduce radical generation, as effective peroxide stabiliser, as preservative potentiator improving piocide efficacy and excellent transition metal chelator.