Learn More
Medchemexpress LLC (S,S)-N,N'-(1,2-dimethyl-1,2-ethanediyl)bis[2,6-piperazinedione] | 21416-88-6 | MFCD01702964 | 99.6% | 282.30 g/mol | C12H18N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11859025MG
Encompass_Preferred
ICRF-193 is a catalytic inhibitor of DNA topoisomerase II used in research to study topoisomerase function and cell cycle regulation. The compound is provided as a small-quantity, high-purity research reagent suitable for in vitro experiments.
  • Catalytic inhibitor of DNA topoisomerase II.
  • High purity (≈99.6%).
  • Molecular weight 282.30 g/mol, formula C12H18N4O4.
  • Supplied as a 25 MG package for small-scale experiments.
  • Suitable for in vitro studies of topoisomerase inhibition and cell cycle analysis.
Catalog No. 50-002-28389