Learn More
Strem, An Ascensus Company CAS# [849950-53-4 ]. 2g. (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, min. 98% (S,S)-Ph-BPE-Rh. MFCD07369038
SDP
Supplier:  Strem, An Ascensus Company 4502022G
Encompass
CAS# 849950-53-4. 2g. (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, min. 98% (S,S)-Ph-BPE-Rh. MFCD07369038. Molecular Weight: 804.49. Molecular Formula: [Rh(C8H12)(C34H36P2)]+BF4ae. Color/form: orange xtl. Strem# 45-0202. www.strem.com/catalog/v3/45-0202/
Catalog No. 50-197-9335